missing translation for 'onlineSavingsMsg'
Learn More
(Formylmethylene)triphenylphosphorane, 97%, may cont. up to ca 3% water, Thermo Scientific Chemicals Product Code.: 11498900

(Formylmethylene)triphenylphosphorane, 97%, may cont. up to ca 3% water, Thermo Scientific Chemicals

Product Code. 11498900
100 g
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11498900

Brand: Thermo Scientific Alfa Aesar A11709.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Formylmethylene)triphenylphosphorane is used as Wittig reagent. It is also used as Pharmaceutical Intermediates. It is employed as raw material in organic Synthesis, Agrochemicals.

Solubility
Soluble in Chloroform, Methanol.

Notes
Air Sensitive. Store at -20°C. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2136-75-6
Molecular Formula C20H17OP
Molecular Weight (g/mol) 304.329
MDL Number MFCD00006994
InChI Key CQCAYWAIRTVXIY-UHFFFAOYSA-N
Synonym 2-triphenylphosphoranylidene acetaldehyde, formylmethylene triphenylphosphorane, triphenylphosphoranylidene acetaldehyde, formylmethylenetriphenylphosphorane, acetaldehyde, triphenylphosphoranylidene, 2-triphenyl-??-phosphanylidene acetaldehyde, acmc-209fjg, ksc492c5lShow MoreShow Less
PubChem CID 75051
IUPAC Name 2-(triphenyl-$l^{5}-phosphanylidene)acetaldehyde
SMILES C1=CC=C(C=C1)P(=CC=O)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 183°C to 188°C
Quantity 100 g
Beilstein 523797
Sensitivity Air sensitive
Solubility Information Soluble in Chloroform,Methanol.
Formula Weight 304.33
Percent Purity 97%
Concentration or Composition (by Analyte or Components) May contain up to ≈3% water
Chemical Name or Material (Formylmethylene)triphenylphosphorane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10217180 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA11709.14,A11709.22,A11709.09
Lot Number10241322 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog NumberA11709.14,A11709.22,A11709.09
Lot Number10169502 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11709.14,A11709.22,A11709.09
Lot Number10179083 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11709.14,A11709.22,A11709.09
Lot Number10225924 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11709.14,A11709.22,A11709.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(Formylmethylene)triphenylphosphorane, 97%, may cont. up to ca 3% water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.