missing translation for 'onlineSavingsMsg'
Learn More
Fumonisin B2, 96+%, Thermo Scientific Chemicals Product Code.: 10440931

Fumonisin B2, 96+%, Thermo Scientific Chemicals

Product Code. 10440931
1 mg
Click to view available options
Quantity:
1 mg
This item is not returnable. View return policy

Product Code. 10440931

Brand: Thermo Scientific Acros 328260010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fumonisin B2, CAS # 116355-84-1, is a diester of tricarballylic acid and is isolated from Fusarium.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Fumonisin B2 is a fumonisin mycotoxin produced by the fungus Fusarium
  • Fumonisin B2 is a structural analog of fumonisin B1 but it is more cytotoxic than B1 and can inhibit sphinganine-N-acetyltransferase (ceramide synthase)

Applications

  • In vitro and in vivo studies have shown Fumonisin B2 can induce apoptosis
  • It can inhibit de novo sphingolipid biosynthesis
  • It is also considered an Aspergillus metabolite
  • It shows cytotoxicity against a panel of seven rat hepatoma cell lines, and MDCK cell line
  • It induces hepatocyte nodule formation, a marker of cancer initiation, in rats
TRUSTED_SUSTAINABILITY

Specifications

CAS 116355-84-1
Assay Percent Range 96+%
Molecular Formula C34H59NO14·3H2O
Quantity 1 mg
Synonym fumonisin b2, 2r,2'r-1,2,3-propanetricarboxylic acid 1,1'-1s,2r-1-2s,9r,11s,12s-12-amino-4,9,11-trihydroxy-2-methyltridecyl-2-1r-1-methylpentyl-1,2-ethanediyl ester, 2s-2-2-5r,6r,7s,9s,16r,18s,19s-19-amino-6-3r-3,4-dicarboxybutanoyl oxy-16,18-dihydroxy-5,9-dimethylicosan-7-yl oxy-2-oxoethyl butanedioic acid
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
InChI Key UXDPXZQHTDAXOZ-FWPUDPCBSA-N
SMILES CCCCC(C)C(C(CC(C)CCCCCCC(CC(C(C)N)O)O)OC(=O)CC(CC(=O)O)C(=O)O)OC(=O)CC(CC(=O)O)C(=O)O
IUPAC Name (2S)-2-[2-[(5R,6R,7S,9S,16R,18S,19S)-19-amino-6-[(3R)-3,4-dicarboxybutanoyl]oxy-16,18-dihydroxy-5,9-dimethylicosan-7-yl]oxy-2-oxoethyl]butanedioic acid
Molecular Weight (g/mol) 759.9
PubChem CID 71317007
Formula Weight 759.9
Percent Purity 96+%
Chemical Name or Material Fumonisin B2
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435996 Certificate TypeCertificate of Analysis Date11/11/2024 Catalog Number328260010
Lot NumberA0462329 Certificate TypeCertificate of Analysis Date30/04/2024 Catalog Number328260010
Lot NumberA0412719 Certificate TypeCertificate of Analysis Date16/01/2023 Catalog Number328260010
Lot NumberA0397748 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number328260010
Lot NumberA0412719 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number328260010
compliance-icons compliance-icons
Product Identifier
  • Fumonisin B2
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fumonisin B2, 96+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.