missing translation for 'onlineSavingsMsg'
Learn More
Furfuryl Alcohol, 98%, Thermo Scientific Chemicals Product Code.: 10255440

Furfuryl Alcohol, 98%, Thermo Scientific Chemicals

Product Code. 10255440
1 L, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10255440

Brand: Thermo Scientific Acros 119790010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-00-0
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
MDL Number MFCD00003252
InChI Key XPFVYQJUAUNWIW-UHFFFAOYSA-N
Synonym furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuranShow MoreShow Less
PubChem CID 7361
ChEBI CHEBI:207496
IUPAC Name furan-2-ylmethanol
SMILES C1=COC(=C1)CO

Specifications

Melting Point -29°C
Color Yellow
Density 1.1300g/mL
Boiling Point 170°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4850 to 1.488
Quantity 1 L
Beilstein 17, V,3, 338
Fieser 01,408
Merck Index 15, 4334
Specific Gravity 1.13
Solubility Information Solubility in water: miscible but unstable
Viscosity 5 mPa.s (25°C)
Formula Weight 98.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Furfuryl alcohol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466100 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog Number119790010,119790025,119792500,119790100
Lot NumberA0461821 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog Number119790010,119790025,119792500,119790100
Lot NumberA0458835 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number119790010,119790025,119792500,119790100
Lot NumberA0457445 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number119790010,119790025,119792500,119790100
Lot NumberA0450286 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number119790010,119790025,119792500,119790100
compliance-icons compliance-icons
Product Identifier
  • Furfuryl alcohol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Furfuryl Alcohol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.