missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl Alcohol, 98%, Thermo Scientific Chemicals

£30.95 - £613.05

Chemical Identifiers

CAS 98-00-0
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.1
MDL Number MFCD00003252
InChI Key XPFVYQJUAUNWIW-UHFFFAOYSA-N
Synonym furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
PubChem CID 7361
ChEBI CHEBI:207496
IUPAC Name furan-2-ylmethanol
SMILES C1=COC(=C1)CO
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10072543
View Documents Promotion Details
Thermo Scientific Acros
119792500
250 mL Glass bottle
£30.95
250mL
Please to purchase this item. Need a web account? Register with us today!
10255440
View Documents
Thermo Scientific Acros
119790010
1 L Glass bottle
£79.90
1 L
Please to purchase this item. Need a web account? Register with us today!
10265440
View Documents
Thermo Scientific Acros
119790025
2.5 L Glass bottle
£154.77
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10701155
View Documents
Thermo Scientific Acros
119790100
10 L Plastic drum
£613.05
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-00-0
98.1
XPFVYQJUAUNWIW-UHFFFAOYSA-N
7361
furan-2-ylmethanol
C5H6O2
MFCD00003252
furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
CHEBI:207496
C1=COC(=C1)CO

Specifications

98-00-0
Yellow
170°C
Authentic
C5H6O2
MFCD00003252
01,408
1.13
Solubility in water: miscible but unstable
C1=COC(=C1)CO
98.1
CHEBI:207496
98.1
Liquid
-29°C
1.1300g/mL
65°C
97.5% min. (GC)
1.4850 to 1.488
17, V,3, 338
15, 4334
furfuryl alcohol, 2-furanmethanol, 2-furylmethanol, 2-furancarbinol, 2-furylcarbinol, furfuranol, 2-furanylmethanol, furfural alcohol, 2-furfuryl alcohol, 5-hydroxymethylfuran
XPFVYQJUAUNWIW-UHFFFAOYSA-N
furan-2-ylmethanol
7361
5 mPa.s (25°C)
98%
Furfuryl alcohol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer if inhaled.
May cause damage to organs through prolonged or repeated exposure if inhaled.
Toxic if swallowed.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Imm

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-626-1

Product Identifier
  • Furfuryl alcohol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P362-Take off contaminated clothing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only