Learn More
Carbolite™ Furnace with Uniform Temperature Envelope
Brand:  Carbolite™ CWF1305-230SN+&02-301
 View more versions of this product
Product Code. 12933741
  • £4585.00 / Each
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
Specifications
301 PID Controller
1300°C
250 x 140 x 135 mm
140 mm
135 mm
250 mm
230 V
50/60 Hz
5 L
Furnace
375 mm
585 mm
485 mm
230 V 50/60 Hz 2400 w
2400 w
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Carbolite™ Furnace with Uniform Temperature Envelope > Capacity: 5L; Temperature Max: 1300C