Learn More
Wld Tec™ Fuego Pro SCS Gas Burner
Safe, streamlined, versatile and energy-efficient laboratory gas burner. Wld Tec™ Fuego Pro SCS Gas Burner is fabricated of stainless steel and heat-resistant glass.
Brand:  Wld Tec™ 8.204.000
Product Code. 12695670
  • £897.52 / Each
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Button or foot pedal operation
  • Gas inlet filter
  • DVGW approved
  • Animated graphic display
  • Battery operation option
  • Gas consumption display
  • Modern design
    • Low casing promotes working comfort
    • Reduced air distrubance
    • Removable burner head for cleaning
    • Integrated drain
  • Universal
    • Accepts stationary natural gas and propane / butane gas supplies, cartridge gas, or gas cylinders
  • Safe
    • Overheating protection
    • Continuous burner head control
    • Burner head clogging detection
    • Residual heat display
    • Automatic cut-off function prevents unintentional ignition of the flame when the burner has not been ignited for a longer period
    • Acoustic signals
  • Environmentally conscious
    • Energy efficiency in accordance with the most recent standards
  • Take-back guarantee
    • Older WLD-TEC burners will be taken back and disposed of in an environmentally compatible manner
  • Robust
    • Stainless steel construction extends to the controls and gas connection
    • Heat-resistant glass display
Specifications
Specifications
Laboratory Gas Burner
103 mm
49 mm
1300°C (Natural Gas), 1350°C (Liquid Gas)
700 g
Natural Gas, Butane, Propane
Stainless Steel
Foot Pedal and Button Function
1 Ea.
130 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Wld Tec™ Fuego Pro SCS Gas Burner > Includes: Animated graphic display, Single or Doubleclick IR-Sensor, Battery operation, Cooling reminder, Zero-pressure shutdown, Gas consumption display