missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified

A soluble form of basement membrane extracted from murine Engelbreth-Holm-Swarm (EHS) tumors. It is free of viruses, including lactose dehydrogenase elevating virus (LDEV), sometimes found in this type of preparation.

Brand:  Gibco™ A1413301

 View more versions of this product

Product Code. 11612149

  • £55.50 / 1mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

  • Routinely used for attachment and maintenance of human embryonic stem cells (hESC)
  • Human ESCs have specific needs for attachment and maintenance and some lots of basement membrane extract may not provide adequate support
  • Geltrex™ hESC-qualified reduced growth factor basement membrane matrix has been tested for support of hESC growth and pluripotency, therefore minimizing the need to test multiple lots
  • Consistent manufacturing processes help ensure a protein concentration averaging 15 mg/mL with very little variation from lot-to-lot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Human induced pluripotent stem cells (iPSC), primary epithelial cells, endothelial cells, smooth muscle cells
12 to 18 mg/mL
Basement Membrane Matrix
Sterile
Gibco™
Dry Ice
Tube(s)
Mouse
Adherent Cell Culture
Reduced Growth Factor
Frozen
Serum-free
No Phenol Red
1 mL
Low
Cell Culture Grade
Geltrex™ LDEV-Free hESC-qualified comes as a sterile protein solution in 1 mL vials.

Store at -20° to -80°C.

Geltrex™ Matrix is stable for 18 months from date of manufacture when properly stored.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number963714 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA1413301,A1413302
Lot Number963676 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA1413301,A1413302
Lot Number963678 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA1413301,A1413302
Lot Number963690 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA1413301,A1413302
Lot Number963694 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA1413301,A1413302
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Geltrex™ Reduced-Growth Factor Basement-Membrane Matrix, LDEV-free, stem-cell qualified > 1 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.