missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneBLAzer™ Gqi5-NFAT-bla FreeStyle™ 293F Cells

GeneBLAzer™ Gqi5-NFAT-bla FreeStyle™ 293F cells contain the chimeric G-protein Gqi5 stably integrated into the CellSensor™ NFAT-blaFreeStyle™ 293F cell line.

Brand:  Thermo Scientific™ K1731

Product Code. 10711634

  • £11260.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GeneBLAzer™ Gqi5-NFAT-bla FreeStyle™ 293F cells contain the chimeric G-protein Gqi5 stably integrated into the CellSensor™ NFAT-blaFreeStyle™ 293F cell line. CellSensor™ NFAT-bla FreeStyle™ 293F cells contain a beta-lactamase reporter gene under control of a stably integrated NFAT response element. Gqo5-NFAT-bla FreeStyle™ 293F is a parental cell line designed to be used for the construction of Gi/o-coupled GPCR beta-lactamase cellular assays. The Gi/o receptor of interest should be stably expressed in the cell line by expression from the selectable vector pcDNA™6.2-hygro-DEST (Cat. No. K1233). Gqi5-NFAT-bla FreeStyle™ 293F cells have been shown to respond to the agonist of a transiently expressed D2 receptor.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dividing Cells
Cell Line: FreeStyle™ 293-F Cells
GPCRs
GeneBLAzer™
BLA (Beta-Lactamase)
FreeStyle 293-F Cells
GPCR
Shipping: Dry Ice
Storage: Liquid Nitrogen
Fluorescence
Gqi5
2 x 106 Cells
Cell Line
GeneBLAzer™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number37762 Certificate TypeCertificate of Analysis Date15/04/2017 Catalog NumberK1731
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ GeneBLAzer™ Gqi5-NFAT-bla FreeStyle™ 293F Cells > 2 x 106 Cells

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.