missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Dispase II, powder

Amino-endo peptidase that hydrolyzes N-terminal peptide bonds of non-polar amino acid residues

Brand:  Gibco™ 17105041

451.64 GBP valid until 2025-12-31
Use promo code "24335" to get your promotional price.


Product Code. 11510536

  • £520.00 / 5g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Dispase II has a mild proteolytic action that makes it useful for the isolation and routine passaging of primary cells. Dispase II is relatively gentle, dissociates well at physiological temperature and pH, and generally maintains cell membrane integrity.

  • Produced in Bacillus polymyxa
  • Packaged as lyophilized, non-sterile powder for research use in cell or tissue dissociation, and to prevent clumping in suspension cultures
  • Activity is >0.5 units/mg

Cell Culture, Mammalian Cell Culture

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2°C to 8°C. Protect from light.
Shipping conditions: Room temperature
Shelf life: 24 months from date of manufacture
Cell Culture Dissociation Reagent
5 g
Lyophilized
Non-sterile
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2581593 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number17105041
Lot Number2581594 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog Number17105041
Lot Number2581595 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number17105041
Lot Number2832273 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number17105041
Lot Number2832274 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number17105041
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Dispase II, powder > 5g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.