Learn More
Gibco™ HBSS, no calcium, no magnesium, no phenol red

Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Brand: Gibco™ 14175095
20.64 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation.
We offer a variety of Gibco™ HBSS formulations for a range of cell culture applications.
This HBSS is modified as follows:
With: Glucose
Without: Calcium, Magnesium, Phenol Red
The complete formulation is available.
Order Info
Shipping Condition: Room Temperature

Specifications
No Sodium Pyruvate | |
Storage conditions: 15°C to 30°C Shipping conditions: Room temperature Shelf life: 36 months from date of manufacture |
|
Gibco™ | |
500 mL | |
Mammalian Cell Culture | |
6.7 - 7.8 | |
Room Temperature | |
Liquid |
Clear | |
1 X | |
Hanks' Balanced Salt Solution (HBSS) | |
Hank's Balanced Salt | |
270 - 295 mOsm/kg | |
No Phenol Red | |
Sterile-filtered | |
cGMP-compliant under the ISO 13485 standard |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3062734 | Certificate of Analysis | 31/01/2025 | 14175095 |
3086141 | Certificate of Analysis | 31/01/2025 | 14175095 |
3062427 | Certificate of Analysis | 31/01/2025 | 14175095 |
2660057 | Certificate of Analysis | 29/01/2025 | 14175095 |
2323431 | Certificate of Analysis | 29/01/2025 | 14175095 |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
78Hours
18Minutes
42Seconds
24New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.