Learn More
Gibco™ Medium 199 (10X)

Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells.
Brand: Gibco™ 11825015
Description
Medium 199 was originally developed for nutritional studies of chick embryo fibroblasts. It has broad species applicability, particularly for cultivation of non-transformed cells. Medium 199 is widely used in virology, vaccine production, and in vitro cultivation of primary explants of mouse pancreatic epithelium, and rat lens tissues. We offer a variety of Gibco™ Medium 199 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.
This M199 is modified as follows:
With: Phenol Red
Without: L-glutamine, HEPES, Sodium Bicarbonate
The complete formulation is available.
Compared to other basal media, Medium 199 contains unique components, including adenine, adenosine, hypoxanthine, thymine, and additional vitamins. Medium 199 is available with Earle's salts for use in a CO2 incubator, or with Hanks' salts for use without CO2.
Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
Medium 199 contains no proteins, lipids, or growth factors. Medium 199 uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.
Concentrated forms of Gibco™ Medium 199 require sodium bicarbonate addition, pH adjustment, and filtration at the time of preparation (see protocol for details).
Order Info
Shipping Condition: Room Temperature

Specifications
Rat epithelial | |
Animal Origin | |
Liquid | |
Sterile-filtered | |
No Glutamine, No HEPES, No Sodium Pyruvate, No Sodium Bicarbonate | |
Gibco™ | |
Room Temperature |
Chick Embryo Fibroblasts, Mouse Pancreatic Epithelium, Rat Lens Tissues | |
10X | |
Medium 199 | |
High Glucose, Phenol Red | |
cGMP-compliant under the ISO 13485 standard | |
500 mL | |
Storage conditions: 2-8° C. Protect from light Shipping conditions: Ambient Shelf life: 12 months from date of manufacture |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2678983 | Certificate of Analysis | 29/01/2025 | 11825015 |
2898833 | Certificate of Analysis | 25/01/2025 | 11825015 |
3063890 | Certificate of Analysis | 25/01/2025 | 11825015 |
A0463578 | Certificate of Analysis | 06/06/2024 | 118255000,11825015 |
A0439583 | Certificate of Analysis | 15/01/2024 | 118255000,11825015,118250025 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures. Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law. For in vitro diagnostic use.