missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Sf-900™ II SFM
GREENER_CHOICE

Optimized for the growth and maintenance of Spodoptera frugiperda (Sf9 and Sf21) cells

Brand:  Gibco™ 10902088

114.89 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10227402

  • £129.00 / 1 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Sf-900 II SFM is a serum-free, protein-free insect cell culture medium for large-scale production of recombinant proteins expressed using the baculovirus expression vector system (BEVS). This medium is suitable for suspension and monolayer culture methods and supports growth of other lepidopteran cell lines.

  • Superior long-term, high-density growth
  • Optimized for recombinant protein production
  • Serum-free, protein-free, ready-to-use formulation
  • Scalable in bioreactors

Bioproduction, Cell Culture, Insect Cell Culture, Sf9 and Sf21 Cell Culture, Viral Vaccine Production

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sf21, Sf9
Protein-free, Serum-free
Insect Cell Serum Free Medium (SFM)
Glutamine
1000 mL
S. frugiperda, Spodoptera frugiperda
Insect Cell
Liquid
Serum-free
Gibco™, Sf-900™
Room Temperature
Storage conditions: 2°C to 8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Sf-900™ II SFM > Quantity: 1000 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.