missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ MICROMAN™ E Capillary Pistons
GREENER_CHOICE

MICROMAN™ E Capillary Pistons provide reproducible, reliable results even with the most viscous or volatile liquids.

£57.84 - £725.00

Specifications

Color Clear
Packaging Box
Product Type Capillary Piston
Disposable Single-use
For Use With (Equipment) Gilson Microman
View More Specs

Products 19
Product Code Brand Volume (Metric) Model Format Quantity Price Quantity & Availability  
Product Code Brand Volume (Metric) Model Format Quantity Price Quantity & Availability  
10192852
GREENER_CHOICE
View Documents
Gilson™
F148413
1 to 10 μL CP10ST TIPACK 2 x 96 192 Pk. N/A N/A N/A
10011912
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148313
1 to 10 μL CP10ST TIPACK 10 x 96 960 Pk.
£725.00
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
15306564
GREENER_CHOICE
View Documents
Gilson™
F148712
3 to 25 μL CP25ST Box 576 Pk.
£260.10
Pack of 576
In Stock
to see stock.
Add to basket
 
10789324
GREENER_CHOICE
View Documents
Gilson™
F148415
10 to 100 μL CP100ST Box 192 Pk. N/A N/A N/A
11327920
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148315
10 to 100 μL CP100ST Box 960 Pk.
£725.00
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
15304785
GREENER_CHOICE
View Documents
Gilson™
F148713
20 to 50 μL CP50ST Box 576 Pk. N/A N/A N/A
15314785
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148714
50 to 250 μL CP250ST Box 576 Pk.
£316.00
Pack of 576
In Stock
to see stock.
Edge
Add to basket
 
12609621
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148180
100 to 1000 μL CP1000ST Box 182 Pk.
£151.00
Pack of 182
In Stock
to see stock.
Edge
Add to basket
 
10041932
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148412
1 to 10 μL CP10 TIPACK 2 x 96 960 Pk.
£118.00
Pack of 192
In Stock
to see stock.
Edge
Add to basket
 
12001532
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148312
1 to 10 μL CP10 TIPACK 10 x 96 960 Pk.
£493.00
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
10463942
GREENER_CHOICE
View Documents
Gilson™
F148112
3 to 25 μL CP25 Box 200 Pk. N/A N/A N/A
10726470
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148012
3 to 25 μL CP25 TIPACK 10 x 96 960 Pk.
£545.00
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
10646653
GREENER_CHOICE
View Documents
Gilson™
F148414
10 to 100 μL CP100 Box 192 Pk.
£75.03
Pack of 192
In Stock
to see stock.
Add to basket
 
10494132
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148314
10 to 100 μL CP100 Box 960 Pk.
£493.00
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
12123771
GREENER_CHOICE
View Documents
Gilson™
F148113
20 to 50 μL CP50 Box 200 Pk. N/A N/A N/A
10866510
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148013
20 to 50 μL CP50 TIPACK 10 x 96 960 Pk.
£545.00
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
10163522
GREENER_CHOICE
View Documents
Gilson™
F148114
50 to 250 μL CP250 Box 200 Pk.
£71.03
Pack of 200
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
 
10456925
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148014
50 to 250 μL CP250 Box 960 Pk.
£545.00
Pack of 960
In Stock
to see stock.
Edge
Add to basket
 
12597526
GREENER_CHOICE
View Documents Promotion Details
Gilson™
F148560
100 to 1000 μL CP1000 Box 182 Pk.
£132.00
Pack of 182
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Recommended for reproducible, reliable results even with the most viscous or volatile liquids.
  • Made specifically for Gilson™ MICROMAN™ E pipets.
  • Available in a wide range of volumes and designs, both in sterilized and standard versions, to suit your needs.
Specifications

Specifications

Clear
Capillary Piston
Gilson Microman
Box
Single-use
Polypropylene
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gilson™ MICROMAN™ E Capillary Pistons

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.