missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Glass Fiber Nylon Spatula

Manufactured from glass fiber nylon for excellent chemical resistance. Kartell™ Glass Fiber Nylon Spatulas are suitable for use in food and beverage industry, laboratory work and educational settings.

Brand:  Kartell™ 0059600

Product Code. 10245281

  • £102.86 / Pack of 10
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Manufactured from glass fiber nylon for excellent chemical resistance. Kartell™ Glass Fiber Nylon Spatulas are suitable for use in food and beverage industry, laboratory work and educational settings.

  • General purpose spatulas with excellent chemical resistance
  • Scraper plus spoon or double scraper configuration
  • Autoclavable
  • Manufactured to DIN12890 standard
Specifications

Specifications

10 Pk.
Spoon
Red
For use in food and beverage industry, inexpensive general purpose spatulas
Glass Fiber Nylon
Autoclavable
1.5 mL
NSF/ANSI Standard 51, DIN 12890
13 to 17 mm
180 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kartell™ Glass Fiber Nylon Spatula > Length: 180mm; with Spoon Ends

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.