missing translation for 'onlineSavingsMsg'
Learn More
Labconco™ Glove Box Gloves, Sleeves and Hands Product Code.: 16467130
Add to basket

Labconco™ Glove Box Gloves, Sleeves and Hands

Product Code. 16467130 Shop All Labconco Corporation Products
Butyl, 9.75, 15 mil, Black
Click to view available options
Material:
Butyl
Neoprene
Size:
9.75
Thickness (English):
15 mil
Color:
Black
This item is not returnable. View return policy

Product Code. 16467130

Brand: Labconco™ 5005601

Add to basket
Add to basket
This item is not returnable. View return policy

Glove box gloves, sleeves and hands are available in a variety of sizes, materials and thicknesses to suit your specific application. Choose from neoprene, latex, CSM or butyl.

Glove Box Gloves - One pair/pack, one-piece, seamless, with O-rings

  • Neoprene - Size 9.75
    • 17 mil thickness
  • CSM (Chlorosulfonated polyethylene rubber) - Available in sizes 8.5 or 9.75
    • Also known as Hypalon
    • Resistant to abrasion, ozone and oxidizing chemicals
    • 15 mil thickness
  • Butyl - Available in sizes 8.5 and 9.75
    • Provides high impermeability to water vapor, gases and toxic chemicals
    • Resistant to oxygenated solvents and most oxidizing chemicals
    • 15 mil thickness

Glove Box Sleeves

  • Neoprene Straight Sleeves
    • For use with Neoprene or Latex Hands
    • With O-rings and springs

Glove Box Hands - For use with neoprene straight or accordian sleeves

  • Neoprene available in size 7.5, 9 or 10
    • 17 mil thickness
    • Lightweight and embossed with firm hold finish
  • Latex available in size 7 or 9
    • 9 mil thickness
    • White, surgical-type

Specifications

Color Black
Hand Ambidextrous
Material Butyl
Size 9.75
Sterility Non-sterile
Thickness (English) 15 mil
Manufacturing/QMS Audit Standards ISO 9001
Model 5005601
  • Multiple Manufacturers
    Discover exclusive monthly deals on essential safety products to enhance your lab's protection and compliance.
    Promo Code: 24258

    Expires2025-03-31

  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Labconco™ Glove Box Gloves, Sleeves and Hands >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.