missing translation for 'onlineSavingsMsg'
Learn More
Glycerol, 98+%, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10336040

Glycerol, 98+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 10336040
2.5 L, HDPE plastic bottle
Click to view available options
Quantity:
500 mL
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10336040

Brand: Fisher Chemical G/0600/17

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Glycerol, 98+%, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglynShow MoreShow Less
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
pH 5
Boiling Point 290°C
Packaging HDPE plastic bottle
Quantity 2.5 L
Vapor Density 3.17
Formula Weight 92.09g/mol
Vapor Pressure 0.003mbar 50
Physical Form Very Viscous Liquid
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Days

    29

    Hours

    8

    Minutes

    17

    Seconds

    15
    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    15

    Minutes

    42

    Seconds

    44
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycerol, 98+%, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.