missing translation for 'onlineSavingsMsg'
Learn More
Glycerol, 99+%, pure, synthetic, Thermo Scientific Chemicals Product Code.: 10001841

Glycerol, 99+%, pure, synthetic, Thermo Scientific Chemicals

Product Code. 10001841
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10001841

Brand: Thermo Scientific Acros 332031000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglynShow MoreShow Less
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18.0°C
Density 1.2610g/mL
Boiling Point 290.0°C
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index 1.4716
Quantity 100 mL
Beilstein 01,502
Merck Index 15,452
Specific Gravity 1.261
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform
Viscosity 1069 mPa.s (20°C)
Formula Weight 92.09
Percent Purity 99+%
Grade Pure
Physical Form Viscous Liquid
Chemical Name or Material Glycerol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469302 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number332030100,332031000,332030010,332030025
Lot NumberA0468973 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number332030100,332031000,332030010,332030025
Lot NumberA0465546 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number332030100,332030000,332030010,332031000,332030025
Lot NumberA0457256 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number332030100,332030000,332030010,332031000,332030025
Lot NumberA0462980 Certificate TypeCertificate of Analysis Date23/05/2024 Catalog Number332030100,332030000,332030010,332031000,332030025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycerol, 99+%, pure, synthetic, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.