missing translation for 'onlineSavingsMsg'
Learn More
Glycerol, puriss., Meets analytical specification of Ph. Eur. BP, USP, FCC., Honeywell™ Riedel-de Haen™ Product Code.: 15621480

Glycerol, puriss., Meets analytical specification of Ph. Eur. BP, USP, FCC., Honeywell™ Riedel-de Haen™
BIOPHARMA

Product Code. 15621480
6 x 1 L, Poly Bottle
Click to view available options
Quantity:
4 x 5 L
6 x 1 L
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15621480

Brand: Honeywell-Riedel-de Haen 155236X1L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E422, anhydrous, 99.0-101.0% (alkalimetric)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglynShow MoreShow Less
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Color Colorless
Density 1.2580 to 1.2644g/mL at 20°C, ≥1.257g/mL at 25°C
Boiling Point 290°C
Flash Point 160°C (320°F)
Ignition Residue ≤0.01% (as SO4)
Ignition Point 698°F
Packaging Poly Bottle
Linear Formula HOCH2CH(OH)CH2OH
Refractive Index n20/D 1.474(lit.)
Quantity 6 x 1 L
UN Number NONH for all modes of transport
Beilstein 635685
Vapor Density 3.1 (vs air)
Formula Weight 92.09g/mol
Vapor Pressure 1mmHg (20°C)
Percent Purity 99.0 to 101.0% (calc. on H2O free substance)
Grade puriss.
Physical Form Liquid
Chemical Name or Material Glycerol, Meets analytical specification of Ph. Eur. BP, USP, FCC.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberO005F Certificate TypeCertificate of Analysis Date02/08/2024 Catalog Number15523-6X1L
Lot NumberN300F Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number15523-6X1L
Lot NumberA0457633 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number15523-6X1L,155235000,155231000,15523-1L,15523-4X5L,15523-5L
Lot NumberN0230S Certificate TypeCertificate of Analysis Date05/04/2023 Catalog Number15523-4X5L,15523-5L,15523-6X1L,15523-1L
Lot NumberN0180S Certificate TypeCertificate of Analysis Date05/04/2023 Catalog Number15523-4X5L,15523-5L,15523-6X1L,15523-1L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Glycerol, puriss., Meets analytical specification of Ph. Eur. BP, USP, FCC., Honeywell™ Riedel-de Haen™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.