Learn More
Glycine (White Crystals or Crystalline Powder), Fisher BioReagents™
£45.15 - £374.00
Chemical Identifiers
CAS | 56-40-6 |
---|---|
Molecular Formula | C2H5NO2 |
Molecular Weight (g/mol) | 75.07 |
MDL Number | MFCD00008131 |
InChI Key | DHMQDGOQFOQNFH-UHFFFAOYSA-N |
Synonym | glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine |
PubChem CID | 750 |
ChEBI | CHEBI:15428 |
IUPAC Name | 2-aminoacetic acid |
SMILES | NCC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10467963
|
Fisher BioReagents
BP381-500 |
500 g | Poly Bottle |
£45.15
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10061073
|
Fisher BioReagents
BP381-1 |
1 kg | Poly Bottle |
£86.95
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10754724
|
Fisher BioReagents
BP381-5 |
5 kg | Poly Pail |
£374.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Glycine (White Crystals or Crystalline Powder)
Glycine is used in the preparation of Tris–Glycine electrophoresis buffers and culture media.Chemical Identifiers
56-40-6 | |
75.07 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
750 | |
2-aminoacetic acid |
C2H5NO2 | |
MFCD00008131 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
CHEBI:15428 | |
NCC(O)=O |
Specifications
182°C | |
0.02% max. | |
1ppm max. | |
White | |
10ppm max. | |
Pass Test | |
98.5 to 101.0 % | |
Pass Test | |
MFCD00008131 | |
glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine | |
1.595g/cm³ | |
2-aminoacetic acid | |
750 | |
75.07 | |
Powder |
0.1 AU max. at 280nm | |
56-40-6 | |
5.9 to 6.4 | |
0.003% max. | |
0.10% max. | |
7ppm max. | |
0.20% max. | |
C2H5NO2 | |
15, 4526 | |
DHMQDGOQFOQNFH-UHFFFAOYSA-N | |
NCC(O)=O | |
75.07 | |
CHEBI:15428 | |
98.5 to 101.0% | |
Glycine |
Safety and Handling
CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:1
Flammability:1
Instability:1
Recommended Storage : RT
Your input is important to us. Please complete this form to provide feedback related to the content on this product.