Learn More
Invitrogen™ Goat anti-Human IgD Secondary Antibody, PE
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ PA186073
386.68 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Description
The preparation has been cross-adsorbed against human IgA, IgG, and IgM to ensure class specificity. May also cross-react with mouse IgD.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Human IgD | |
Polyclonal | |
PE | |
Goat | |
Affinity chromatography | |
RUO | |
Human | |
Secondary Antibody | |
IgG |
Flow Cytometry | |
0.5 mg/mL | |
PBS with proprietary stabilizer and <0.1% sodium azide | |
Human IgD. | |
500 μg | |
Secondary | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.