missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rabbit IgG (H+L) Secondary Antibody, Cy3 (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB120-6939
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Fluorophore-linked immunosorbent assay.Specifications
IgG (H+L) | |
Polyclonal | |
Cyanine3 | |
IGHG | |
Full length native Rabbit IgG (purified). | |
0.1 mg | |
Adaptive Immunity, Immunology | |
This antibody was pre-adsorbed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat, and Sheep Serum Proteins. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Goat Serum, Rabbit IgG and Rabbit Serum. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Mouse, Rat and Sheep Serum Proteins. | |
Rabbit | |
IgG |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), ELISA | |
1 mg/mL | |
Western Blot, Flow Cytometry 1:500 - 1:2500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Immunohistochemistry-Frozen, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
Goat | |
Affinity Purified | |
RUO | |
Secondary | |
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concetration. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Goat anti-Rabbit IgG (H+L) Secondary Antibody, Cy3 (Pre-adsorbed), Novus Biologicals™ > 0.1mg; Cy3
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction