Learn More
GPR3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20847-1-AP-150UL
 View more versions of this product
Product Code. 16820425
  • £349.00 / 150µL
Explore more special offers
Description
Description
The G-protein-coupled receptor (GPCR) superfamily is comprised of an estimated 600-1, 000 members and is the largest known class of molecular targets with proven therapeutic value. GPR3 is an orphan receptor with constitutive G signaling activity. GPR3 modulates a number of brain functions, including behavioral responses to stress, neurite outgrowth and amyloid-beta peptide generation suggesting that it may play a role in Alzheimer′s disease.
Specifications
Specifications
GPR3
Polyclonal
Unconjugated
GPR3
ACCA, ACCA orphan receptor, G protein coupled receptor 3, GPR3
Rabbit
Antigen Affinity Chromatography
RUO
2827
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P46089
GPR3
GPR3 Fusion Protein Ag14257
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
GPR3 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated