Learn More
SPEX Certiprep™ Stainless Steel Grinding Vial Set
Used to grind hard samples like bone and circuit boards
Brand:  SPEX Certiprep™ 6781S
Product Code. 12584316
  • £570.00 / 1 set
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use when contact with plastic is not recommended.
  • Set includes stainless steel impactor, stainless steel center cylinder, two end plugs, and a pack of 10 O-rings.
  • Sample grinding capacity of 0.1 gram - 5.0 gram.
Specifications
Specifications
Freezer Mills
Stainless Steel
0.1 to 0.5 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SPEX Certiprep™ Stainless Steel Grinding Vial Set >