missing translation for 'onlineSavingsMsg'
Learn More

Cytiva GSTrap™ HP Columns

Use to provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins. Cytiva Life Sciences™ GSTrap™ HP Columns are prepacked with Glutathione Sepharose™ High Performance medium.

Brand:  Cytiva 17-5282-02

Code : GE

Additional Details : Weight : 2.14100kg


 View more versions of this product

Product Code. 11565035

  • £1227.00 / Pack of 5
Estimated Shipment: 05-08-2024
to see stock.
Add to basket

Description

Description

  • One-step purification of GST-tagged proteins and other glutathione S-transferases or glutathione-dependent proteins
  • Glutathione coupled to Sepharose™ High Performance media
  • Column material: polypropylene
  • May be used with syringe, peristaltic pump or chromatography system
  • Purification process preserves antigenicity and function
  • Available as 1mL and 5mL columns
Media
  • Average particle size: 34μm
  • Ligand: Glutathione
  • Matrix: Highly cross-linked 6% agarose
  • Binding capacity: >7mg/mL
  • pH stability working range: 3 to 12

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.
5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

Specifications

Specifications

5 mL
16 mm
<20 mL/min.
3 to 12
4°C to 30°C, 20% Ethanol
34 μm
Glutathione S-transferase (GST) tagged proteins produced using the pGEX series of expression vectors, Other glutathione S-transferases and glutathione-dependent proteins
Glutathione
0.62 in.
GSTrap HP columns are prepacked with Glutathione Sepharose High Performance resin and provide high-resolution, one-step purifications of glutathione S-Transferase (GST) tagged proteins.
Polypropylene
4% Highly Cross-linked Agarose
5 bar (0.5 MPa, 70 psi)
5 x 5 mL
Pre-packed Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • GSTrap HP 5 ml, 5 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers