Learn More
Hamilton™ CTC and LEAP Technologies GC PAL Autosampler Syringes: Gastight
Designed for use in PAL GC-xt, PAL COMBI-xt, PAL COMBI-xt extended and other autosampler systems
Brand:  Hamilton™ 203226
 View more versions of this product
Product Code. 10244633
  • £225.00 / Each
Explore more special offers
Description
Show More
Specifications
Specifications
Not Autoclavable
0.057 in.
1.46 mm
Custom
Custom
68°F
20°C
100 μL
PAL GC-xt, PAL COMBI-xt, PAL COMBI-xt extended and other autosampler systems
23 G
0.26 in.
6.6 mm
2 in.
Gastight Syringe
1710 SN
Ea.
Cone
Cemented
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hamilton™ CTC and LEAP Technologies GC PAL Autosampler Syringes: Gastight > Gastight; 100μL; Termination: SN; Custom gauge; Point style: Custom