missing translation for 'onlineSavingsMsg'
Learn More
Hammond™ Desiccant, Cole-Parmer™ Product Code.: 11705599
compliance-icons

Hammond™ Desiccant, Cole-Parmer™

Product Code. 11705599
Non-Indicating Drierite Desiccant, 1 lb.
Click to view available options
Chemical Name or Material:
Indicating Drierite Desiccant
Non-Indicating Drierite Desiccant
Quantity:
1 lb.
5 lb.
This item is not returnable. View return policy

Product Code. 11705599

Brand: Cole Parmer 0719320

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Choose indicating or non-indicating desiccant—indicating desiccant starts blue, then turns pink when regeneration or renewal is necessary. Use Hammond™; Drierite™ Desiccant (anhydrous calcium sulfate) in desiccators, drying columns or drying tubes.

  • Indicating desiccant drying capacity is 10 to 14% of its weight for water and gases 
  • Non-indicating desiccant drying capacity is 6% of its weight for water and gases

Specifications

Material Calcium Sulfate
Quantity 1 lb.
Suitability Used in Desiccators, Drying Columns, or Drying Tubes
Chemical Name or Material Non-Indicating Drierite Desiccant
compliance-icons
Product Identifier
  • drierite
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : crystaline silica
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hammond™ Desiccant, Cole-Parmer™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.