Learn More
Invitrogen™ HCS CellMask™ Stains
Easily visualize entire cells or individual cell structures during high-content screening (HCS) assays with HCS CellMask stains, which are available in a variety of colors for multiplexing flexibility and can be used immediately after fixation and permeabilization, or after antibody labeling.
£347.00 - £386.00
Specifications
For Use With (Equipment) | High Content Instrument |
---|---|
Quantity | 1 Set |
Detection Method | Fluorescence |
Product Line | CellMask™ |
Shipping Condition | Room Temperature |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10555854
|
Invitrogen™
H32720 |
HCS CellMask Blue Stain |
£347.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10266984
|
Invitrogen™
H32714 |
HCS CellMask Green Stain |
£386.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16481821
|
Invitrogen™
H32722 |
HCS CellMask Near-IR Stain |
£370.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10033484
|
Invitrogen™
H32713 |
HCS CellMask Orange Stain |
£352.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10615695
|
Invitrogen™
H32712 |
HCS CellMask™ Red Stain |
£381.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10707065
|
Invitrogen™
H32721 |
HCS CellMask Deep Red Stain |
£381.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Identify entire cells (cytoplasm and nucleus) during high-content screening (HCS) assays with the versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains, which are fluorophores that can be used immediately after fixation and permeabilization, or after antibody labeling. They are available in a variety of colors for multiplexing protocols.
The versatile HCS CellMask Red, CellMask Orange, CellMask Green, CellMask Blue, CellMask Deep Red, and CellMask Near-IR stains are cell delineation tools for high content screening (HCS) platforms, label the entire cell (cytoplasm and nucleus) and provide an accurate backdrop against which the features of interest can be assessed. The HCS CellMask Red and Blue stains replace the HCS CellMask Red cytoplasmic/nuclear (Cat. No. H32711) and HCS CellMask Blue cytoplasmic/nuclear stains (Cat. No. H34558), respectively.HCS CellMask stains can be applied to cells immediately after fixation or in the last step of multiplexing protocols, and they are compatible with detergent-based cell permeabilization protocols. If only nuclear staining is desired, HCS NuclearMask stains can be used for measuring DNA content in addition to enabling robust cell demarcation. HCS NuclearMask stains are also available in a choice of colors for multiplexing flexibility.
Specifications
High Content Instrument | |
Fluorescence | |
Room Temperature | |
Stain |
1 Set | |
CellMask™ | |
Fluorescent Dye | |
Cytoplasm, Cytosol, Nucleus |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.