missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buerkle™ HDPE Wide Neck Square Reagent Bottles
The Wide-neck reagent bottle is particularly suited for storing, packing, taking samples, sampling and transport of granules and paste-like media.
£2.50 - £16.10
Specifications
Color | Transparent |
---|---|
Material | HDPE |
Closure Material | Polypropylene |
Closure Type | Screw Cap |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15475574
|
Buerkle™
0342-0050 |
50 mL |
£2.50
Each |
In Stock
Log in to see stock. |
|||||
15425574
|
Buerkle™
0342-0100 |
100 mL |
£6.35
Each |
In Stock
Log in to see stock. |
|||||
10222563
|
Buerkle™
0342-2500 |
2500 mL |
£9.75
Each |
In Stock
Log in to see stock. |
|||||
17250574
|
Buerkle™
0342-0750 |
750 mL |
£4.55
Each |
In Stock
Log in to see stock. |
|||||
15376418
|
Buerkle™
0342-0250 |
250 mL |
£7.90
Each |
In Stock
Log in to see stock. |
|||||
11817702
|
Buerkle™
0342-0500 |
500 mL |
£8.45
Each |
In Stock
Log in to see stock. |
|||||
15316438
|
Buerkle™
0342-1000 |
1000 mL |
£6.70
Each |
In Stock
Log in to see stock. |
|||||
11868301
|
Buerkle™
0342-1500 |
1500 mL |
£10.20
Each |
In Stock
Log in to see stock. |
|||||
15425584
|
Buerkle™
0342-4000 |
4000 mL |
£16.10
Each |
In Stock
Log in to see stock. |
|||||
Description
- HDPE, transparent
- PP tamper-evident screw cap, blue
- UN approval for bulk goods
Specifications
Transparent | |
HDPE | |
Polypropylene | |
Screw Cap |