missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VELP Scientifica™ Hemispheric Bowls for Round Bottom Flasks
The Hemispheric Bowl is a suitable solution for replacing oil baths and mantles in order to perform clean and safe analysis with round bottom flasks.
Brand: VELP Scientifica™ A00000374
This item is not returnable.
View return policy
Description
- Precisely fits 135 mm diameter plate
- Accommodates different sizes of round bottom flasks
- Minimizes risk of movement
- Ensures fastest heat transfer
Specifications
Round Bottom Flasks | |
3 L | |
Aluminum |
Flask Bowl | |
Aluminum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VELP Scientifica™ Hemispheric Bowls for Round Bottom Flasks > Hemispheric Bowls for Round Bottom Flasks; Vial Volume (Metric): 3 L; Material: Aluminum; Coating: Aluminum; For Use With (Equipment): Round Bottom Flasks; Type: Flask Bowl
Spot an opportunity for improvement?Share a Content Correction