missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™
Commonly used buffering agent.
Brand: Fisher BioReagents BP310-500
184.83 GBP valid until 2025-12-31
Use promo code "24341" to get your promotional price.
This item is not returnable.
View return policy
Description
Commonly used buffering agent
HEPES is commonly used as a buffering agent.
Specifications
0.01 max. (0.1M solution) at 280nm | |
7365-45-9 | |
5.0 to 6.5 | |
500 g | |
DNase-, RNase- and Protease-Free | |
Pass Test | |
MFCD00006158 | |
Protease free | |
N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid | |
JKMHFZQWWAIEOD-UHFFFAOYSA-N | |
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid | |
23831 | |
≥99% |
HEPES | |
White | |
Powder or Solid | |
DNase free | |
≥99 % | |
C8H18N2O4S | |
Poly Bottle | |
15, 4689 | |
Soluble in water | |
OCCN1CCN(CCS(O)(=O)=O)CC1 | |
238.30 | |
CHEBI:42334 | |
Molecular Biology |
Safety and Handling
Product Identifier
- HEPES
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
HEPES (Fine White Crystals/Molecular Biology), Fisher BioReagents™ > Poly Bottle; 500g
Spot an opportunity for improvement?Share a Content Correction