missing translation for 'onlineSavingsMsg'
Learn More
Hexamethyleneimine, 98+%, Thermo Scientific Chemicals Product Code.: 11406916

Hexamethyleneimine, 98+%, Thermo Scientific Chemicals

Product Code. 11406916
100 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11406916

Brand: Thermo Scientific Alfa Aesar B24007.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyleneimine is used as a pharmaceutical product and raw material for rubber products. It is also used as a intermediate for agrochemicals, zeolites, dyes , inks, rubber chemicals, textile chemicals, corrosion inhibitors, ore flotation.

Solubility
Fully miscible in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-49-9
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00006934
InChI Key ZSIQJIWKELUFRJ-UHFFFAOYSA-N
Synonym hexamethyleneimine, homopiperidine, hexahydroazepine, hexahydro-1h-azepine, perhydroazepine, 1h-azepine, hexahydro, hexamethylenimine, azacycloheptane, azepan, hexamethylene imineShow MoreShow Less
PubChem CID 8119
ChEBI CHEBI:32616
IUPAC Name azepane
SMILES C1CCCNCC1

Specifications

Melting Point -37°C
Density 0.88
Boiling Point 138°C to 139°C
Flash Point 22°C (71°F)
Odor Ammonia-like
Refractive Index 1.466
Quantity 100 mL
UN Number UN2493
Beilstein 1084
Solubility Information Fully miscible in water.
Formula Weight 99.18
Percent Purity ≥98%
Chemical Name or Material Hexamethyleneimine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10229055 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24007.AP,B24007.AE
Lot NumberFA031887 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24007.AP,B24007.AE
Lot Number9001 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24007.AP,B24007.AE
Lot Number10115619 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24007.AP,B24007.AE
Lot NumberI20X024 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB24007.AP,B24007.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hexamethyleneimine, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.