Learn More
Thermo Scientific™ High Performance Microtube with Snap-on Seal Closure
Recommended when using microcentrifuge tubes in Thermo Scientific micro-ultracentrifuge rotors.
Brand:  Thermo Scientific™ 314352H01
147.35 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 15361379
  • £155.11 / Pack of 100
Explore more special offers
Description
Description
Recommended when using microcentrifuge tubes in Thermo Scientific micro-ultracentrifuge rotors.
  • 1.5 mL High Performance Microtube with Snap-on Seal Closure.
  • Recommended when using microcentrifuge tubes in Thermo Scientific micro-ultracentrifuge rotors.
  • The maximum RCF of this microtube is 201,000 x g
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Microcentrifuge Tube
1.5 mL
55,000 rpm
42 mm
S110-AT, S100-AT4, S100-AT5, S100-AT6, and S80-AT3 Rotors with Adapters. S55-A2 and S45-A Rotor Are Used Without Adapters.
201,000 x g
11 mm
100
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ High Performance Microtube with Snap-on Seal Closure > S110-AT, S100-AT4, S100-AT5, S100-AT6, and S80-AT3 rotors with adapters. S55-A2 and S45-A rotor are used without adapters.