Learn More
Cytiva His GraviTrap™ TALON™
Offers a different selectivity compared to nickel-charged media
Brand: Cytiva 29000594
Description
Prepacked gravity flow column for convenient manual purification of histidine-tagged proteins
Specificatios:
- Ligand - TALON Superflow Cobalt-IMAC
- Matrix - Cross-linked agarose, 6%
- Particle size - 60μm-160μm
- Binding Capacity/mL - Up to 20 mg histidine-tagged protein
- pH stability working range - 3 to 12
- Column i.d. - 15mm
- Material [Column Hardware] - Polypropylene (PP)
- Material [Bed Support] - Polyethylene (PE)
- Complete packsize - 10
- Bed volume - 1mL TALON Superflow /column

Specifications
10 mL | |
For Convenient Manual PurifiCatIon of Histidine-Tagged Proteins | |
Up to 15 mg Histidine-tagged Protein | |
His GraviTrap Talon prepacked Column | |
60 to 160 μm | |
1 mL |
Stable in commonly used Buffer systems | |
4°C to 8°C | |
0.59 in. | |
1.5 cm | |
6% Cross-Linked Agarose | |
Talon Superflow Cobalt-IMAC |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Warning
Hazard Category
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol, ethanol
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
8Hours
19Minutes
19Seconds
19 -
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
39Hours
19Minutes
19Seconds
19New Offer Save Up to 70%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.