missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Chelating HP Columns

Optimize histidine-tagged protein purifications when nickel ions are not the best choice. Cytiva Life Sciences™ HiTrap™ Chelating HP Columns is prepacked with Chelating Sepharose™ High Performance.

Brand:  Cytiva 17-0409-03

Code : GE

Additional Details : Weight : 0.17400kg


 View more versions of this product

Product Code. 11594895

  • £653.00 / Pack of 5
Estimated Shipment: 06-08-2024
to see stock.
Add to basket

Description

Description

  • Uses different kinds of metal ions
  • Binding capacity ∽12mg histidine-tagged protein/mL medium
  • Metal ion capacity ∽23μmol Cu2+/mL medium
  • Prepacked with Chelating Sepharose High Performance media
  • Iminoacetic acid coupled to a 6% agarose matrix by ether linkage

Specifications

Specifications

6M Guanidine Hydrochloride, 8M Urea
5mL
16mm
0.98 in.
<20mL/min.
Iminodiacetic Acid
3 to 13
6% Highly Cross-linked Agarose
Affinity Column
HiTrap chelating HP column
0.62 in.
5 x 5mL
25mm
Polypropylene
3 bar
34μm
4° to 30°C
IMAC and subsequent purification of Polyhistidine tagged Proteins
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HiTrap Chelating HP 5 ml, 5 x 5 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers