missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Chelating HP Columns
Optimize histidine-tagged protein purifications when nickel ions are not the best choice. Cytiva Life Sciences™ HiTrap™ Chelating HP Columns is prepacked with Chelating Sepharose™ High Performance.
£168.00 - £8571.00
Specifications
Buffer | 6M Guanidine Hydrochloride, 8M Urea |
---|---|
Product Type | HiTrap chelating HP column |
Height (English) | 0.98 in. |
Height (Metric) | 25mm |
Ligand Type | Iminodiacetic Acid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10773017
|
Cytiva
17-0409-01 |
1 x 5mL |
£168.00
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
10605575
|
Cytiva
17-0408-01 |
5 x 1mL |
£193.00
Pack of 5 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
11594895
|
Cytiva
17-0409-03 |
5 x 5mL |
£653.00
Pack of 5 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||||
11850073
|
Cytiva
17-0409-05 |
100 x 5mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Uses different kinds of metal ions
- Binding capacity ∽12mg histidine-tagged protein/mL medium
- Metal ion capacity ∽23μmol Cu2+/mL medium
- Prepacked with Chelating Sepharose High Performance media
- Iminoacetic acid coupled to a 6% agarose matrix by ether linkage
Specifications
6M Guanidine Hydrochloride, 8M Urea | |
0.98 in. | |
Iminodiacetic Acid | |
3 to 13 | |
6% Highly Cross-linked Agarose | |
Affinity Column |
HiTrap chelating HP column | |
25mm | |
3 bar | |
34μm | |
4° to 30°C | |
IMAC and subsequent purification of Polyhistidine tagged Proteins |