missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ NHS-Activated HP Columns

Couple ligands containing primary amines easily. Cytiva Life Sciences™ HiTrap™ NHS-Activated HP Columns are prepacked with NHS activated Sepharose™ High Performance.

Brand:  Cytiva 17-0717-01


 View more versions of this product

Product Code. 11708478

  • £186.00 / 5mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Complete instructions for use and method optimization, plus a variety of connectors for syringe, peristaltic pump, and chromatography systems
This item is not returnable. View return policy

Description

Description

Prepacked with NHS-activated Sepharose™ High Performance Media for coupling ligand via primary amines at close to neutral pH

  • Designed for use with a syringe, peristaltic pump, or chromatography system
  • Degree of substitution: 10 μmol NHS/mL medium
  • Delivered in 100% isopropanol to protect the active groups
  • Stable in the pH range 3 to 12, depending upon ligand
  • Average particle size: 34μm

Open in Edit Popup
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 x 5mL
34 μm
5mL
1.6cm
0.98 in.
10μM NHS groups/mL medium
42psi
6% Highly Cross-linked Agarose
20 mL/min.
Syringe, peristaltic pump or liquid chromato graphy system such as AKTA™ design or FPLC system
NHS-Activated HP Column
Preparative affinity chromato graphy, easy coupling of ligands containing primary Amines
2.5cm
Polypropylene
3 to 12
4° to 8°C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.