missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HiTrap™ Protein L Prepacked Columns

For capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs

Brand:  Cytiva 17-5478-55

Code : GE

Additional Details : Weight : 0.20000kg


 View more versions of this product

Product Code. 12684883

  • £3074.00 / Pack of 5
Estimated Shipment: 07-08-2024
to see stock.
Add to basket

Description

Description

  • Prepacked with Capto L affinity chromatography medium (resin)
  • Polypropylene column are easily connected in series for increased productivity
  • Easy screening and convenient process development
  • For use with syringe, peristaltic pump, or chromatography system
  • Maximum pressure: 5 bar
  • 1 and 5mL columns

BioProcess Media
  • Ligand: Recombinant protein L (E. coli), mammalian free
  • Matrix: Rigid, highly cross-linked agarose
  • Average particle size: 85μm
  • pH stability working range: 2 to 10

1mL columns

  • Column I.D.: 7mm
  • Flow velocity: <500cm/h3

5mL columns
  • Column I.D.: 16mm
  • Flow velocity: <500cm/h4

Specifications

Specifications

HiTrap Protein L column
0.62 in.
5 x 5mL
20mm
Polypropylene
5 bar
85μm
2° to 8°C
Affinity chromato graphy medium (Resin) for capture of Antibodies and Antibody fragments such as Fab fragments, scFv and Dabs
5mL
16mm
0.78 in.
<500cm/hr.
Recombinant Protein L (E. coli), mammalian free
2 to 10
Highly Cross-linked Agarose
Affinity Column
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • hitrap protein L

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P235-Keep cool.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers