missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HiTrap™ Protein L Prepacked Columns
For capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs
£201.00 - £3074.00
Specifications
Product Type | HiTrap Protein L column |
---|---|
Height (English) | 0.78 in. |
Height (Metric) | 20mm |
Flow Rate | <500cm/hr. |
Material | Polypropylene |
Product Code | Brand | Diameter (English) Inner | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Inner | Quantity | Price | Quantity & Availability | ||||
12671206
|
Cytiva
17-5478-15 |
0.62 in. | 1 x 5mL |
£754.00
Each |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
12684883
|
Cytiva
17-5478-55 |
0.62 in. | 5 x 5mL |
£3074.00
Pack of 5 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
||||
12681206
|
Cytiva
17-5478-51 |
0.27 in. | 5 x 1mL |
£784.00
Pack of 5 |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
15149945
|
Cytiva
29048665 |
0.27 in. | 1 x 1mL |
£201.00
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
Description
- Prepacked with Capto L affinity chromatography medium (resin)
- Polypropylene column are easily connected in series for increased productivity
- Easy screening and convenient process development
- For use with syringe, peristaltic pump, or chromatography system
- Maximum pressure: 5 bar
- 1 and 5mL columns
BioProcess Media
- Ligand: Recombinant protein L (E. coli), mammalian free
- Matrix: Rigid, highly cross-linked agarose
- Average particle size: 85μm
- pH stability working range: 2 to 10
1mL columns
- Column I.D.: 7mm
- Flow velocity: <500cm/h3
5mL columns
- Column I.D.: 16mm
- Flow velocity: <500cm/h4
Specifications
HiTrap Protein L column | |
20mm | |
Polypropylene | |
5 bar | |
85μm | |
2° to 8°C | |
Affinity chromato graphy medium (Resin) for capture of Antibodies and Antibody fragments such as Fab fragments, scFv and Dabs |
0.78 in. | |
<500cm/hr. | |
Recombinant Protein L (E. coli), mammalian free | |
2 to 10 | |
Highly Cross-linked Agarose | |
Affinity Column |