Learn More
Hoefer™ Blotting Paper
Cellulose papers precut for convenience
Brand:  Hoefer™ TE46
 View more versions of this product
Product Code. 12014757
  • £64.90 / Pack of 50
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For Western Blotting
Specifications
Specifications
14.5 cm
TE42 or TE62 Tank Blotting Units
Cellulose
21.5 cm
Sheet