Learn More
Hoefer™ Blotting Paper
Cellulose papers precut for convenience
£55.18 - £87.42
Specifications
Format Sheet
Material Cellulose
View More Specs
Products 7
Product Code Brand Length (Metric) Width (Metric) For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) For Use With (Equipment) Price Quantity & Availability  
12014757
View Documents
Hoefer™
TE46
14.5 cm 21.5 cm TE42 or TE62 Tank Blotting Units
£64.90
Pack of 50
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
10331575
View Documents
Hoefer™
TE46
14.5 cm 21.5 cm TE42 or TE62 Tank Blotting Units
£87.42
Pack of 50
In Stock
to see stock.
Add to basket
11544434
View Documents
Hoefer™
TE26
9 cm 10.5 cm TE22 Mini Tank Blotting Unit or MiniVE Blot Module
£65.70
Pack of 50
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
10762097
View Documents Promotion Details
Hoefer™
TE76
21 cm 25 cm TE77 Semi-Dry Blotting Units
£66.87
Pack of 25
In Stock
to see stock.
Add to basket
12069847
View Documents
Hoefer™
TE76
21 cm 25 cm TE77 Semi-Dry Blotting Units
£64.15
Pack of 25
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
10136664
View Documents
Hoefer™
TE76-1416
14 cm 16 cm TE70 or TE77 Semi-Dry Blotting Units This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11544454
View Documents
Hoefer™
TE76-1416
14 cm 16 cm TE70 or TE77 Semi-Dry Blotting Units
£55.40
Pack of 25
Estimated Shipment: 04-02-2025
to see stock.
Add to basket
Description
Description
  • For Western Blotting
Specifications
Specifications
Sheet
Cellulose