Learn More
Hoefer™ Blotting Paper
Cellulose papers precut for convenience
Brand:  Hoefer™ TE76
 View more versions of this product
Product Code. 12069847
  • £64.15 / Pack of 25
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For Western Blotting
Specifications
Specifications
21 cm
TE77 Semi-Dry Blotting Units
Cellulose
25 cm
Sheet