Learn More
Hoefer™ Hoefer™ Multiple Gel Caster
Enables several gradient or homogenous gels to be cast simultaneously. Cast homogenous or gradient gels.
Brand:  Hoefer™ SE615
Product Code. 12064707
  • £941.00 / Each
Explore more special offers
Description
Description
  • Casts up to ten 16cm long gels
  • Easy to use and leak free
  • Cast homogenous or gradient gels
    • Pour homogenous gels directly into the top of the casting chamber
    • Pump gradient gels through an inlet port at the bottom
Includes:
  • Caster Body with Gasket
  • Spacer-Mate–Alignment Template
  • Face Plate
  • Thumbscrews–2 pcs
  • Clamps–4 pcs
  • Filler Plug Set
  • Glass Plates
  • Black Vinyl Cap
  • Filter Sheets–5 pcs
  • Gel Seal
  • Wax Paper–100 Sheets, Precut
  • Bubble Level
Specifications
Specifications
Ten Gel Caster
Multiple Gel Caster
Includes 20 Glass Plates, 18 x 16cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Hoefer™ Hoefer™ Multiple Gel Caster > For Use With: 18 x 16cm Plates Size