Learn More
Thermo Scientific™ HotStart™ Storage Reaction Tubes
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Brand: Thermo Scientific™ 6002-11
Description
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Features:- Bead is stable at room temperature
- Assures synchronous reaction startup
- PCR product can remain in tube for later analysis
- Hinged caps provide snug seal
- Presterilized; RNase- and DNase-free
Specifications
Hot Start 50 | |
25 to 50 μL | |
0.5 mL | |
Tube |