Learn More
R&D Systems™ Human GDNF Quantikine ELISA Kit
Human GDNF Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Brand: R&D Systems™ DGD00
440.80 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
The Human GDNF Quantikine ELISA Kit quantifies human GDNF in cell culture supernates (50 μL), cell lysates (50 μL), tissue lysates (50 μL), serum (50 μL), edta plasma (50 μL), heparin plasma (50 μL).
Specifications
4.69 to 300 pg/mL | |
Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Tissue Lysates | |
2668 | |
1 Kit | |
Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
0.696 pg/mL | |
Natural and Recombinant Mouse ELA2 | |
Human | |
Cell Culture Supernates (50 μL),Cell Lysates (50 μL),Tissue Lysates (50 μL),Serum (50 μL),EDTA Plasma (50 μL),Heparin Plasma (50 μL) | |
4 hr. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
R&D Systems™ Human GDNF Quantikine ELISA Kit > 1 Kit
Spot an opportunity for improvement?Share a Content Correction