Learn More
Proteintech HumanKine™ recombinant human EGF protein
HumanKine™ recombinant protein
Brand:  Proteintech HZ-1326-10UG
 View more versions of this product
Product Code. 17620107
  • £37.00 / 10µg
Explore more special offers
Specifications
P01133
1950
10 μg
Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Human
Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Lyophilized
1 x PBS, See Certificate of Analysis for details
7 kDa, monomer, glycosylated
HEK293
<1.0 EU/μg
Recombinant
0.1-0.6 ng/mL EC50
>95%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Proteintech HumanKine™ recombinant human EGF protein > 10 μg