missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyCell™ TransFx™ H Media

Brand:  Cytiva SH30939.02

93.99 GBP valid until 2025-12-31
Use promo code "24275" to get your promotional price.


Product Code. 15280496

  • £141.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

HyCell TransFx-H media for transient transfection of HEK293 cells and production of a variety of recombinant proteins manufactured under cGMP in ISO-compliant facilities.

Highlights:

  • Defined hydrolysate-free ADCF cell culture media formulation
  • Supports transient transfection and growth of multiple HEK293 cell lines
  • Supports the transient expression of a variety of recombinant proteins
  • Same media for transient transfection and production
  • Developed through our Metabolic Pathway Design process for high transfection efficiency, high viable cell yields, and recombinant protein production
  • Component traceability
  • Manufactured under cGMP in ISO-compliant facility

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2° to 8°C, away from light
HyCell TransFx-H liquid medium - 1000 mL
1000 mL
Liquid
0.2 μm Sterile-filtered
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyCell™ TransFx™ H Media > 1L PETE bottle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.