missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Gas Delivery Tube

Polypropylene construction. Azlon™ Gas Delivery Tube are ideal substitutes for fragile glass alternatives.

Brand:  Azlon™ DT003

 View more versions of this product

Product Code. 12683012

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Pre-bent rigid polypropylene delivery tubes
  • Unbreakable in normal use
  • Suitable for many types of gas collection experiments in schools and universities
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene
13.18 in.
Many types of gas collection experiments in schools and universities
Side Arm Gas Delivery Tube
6 mm
335 mm
135°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azlon™ Gas Delivery Tube > Double bent; Side arm; Length: 335mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.