missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Cosmic Calf™ Serum, EMEA Tested
Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA)
£169.00 - £178.50
Specifications
Product Type | Calf Bovine Serum |
---|---|
Quantity | 1000 mL |
Sterility | Sterile |
Sterilization Method | Sequential Triple-filtered, 100 nm |
Product Line | HyClone™ |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
16695765
|
Cytiva
SH30533.04 |
Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16605775
|
Cytiva
SH30533.04NI |
Non-Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- High-Quality Bovine Calf Serum
- Fortified with iron and naturally derived components
- Augmented with additional growth-promoting components to yield an exceptional bovine calf serum
- Excellent performance with a variety of cell types
Specifications
Calf Bovine Serum | |
Sterile | |
HyClone™ |
1000 mL | |
Sequential Triple-filtered, 100 nm | |
European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests |