missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ DMEM/HIGH Powdered Media

Supports cell culture growth and biomanufacturing processes with powdered DMEM high glucose media. HyClone™ DMEM/HIGH Powdered Media is manufactured using ISO 9001 processes.

Brand:  Cytiva SH30003.03

 View more versions of this product

Product Code. 11501781

  • £30.15 / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High-quality classical media formulations in convenient packaging
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Exacting manufacturing processes ensure lot–to–lot consistency
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Powder
Non-sterile
No Sodium Pyruvate, No Sodium Bicarbonate
cGMP and ISO 9001:2000
DMEM/High glucose with L-glutamine, without Sodium Pyruvate
Glutamine, Glucose
10 L
2° to 8°C, away from light and moisture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva HyClone™ DMEM/HIGH Powdered Media > Qty: 1 x 10L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.