missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum, US Origin
Contains additional iron that boosts serum transferrin levels to physiological levels
£15.86 - £157.00
Specifications
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities |
---|---|
Age | 16 to 22 weeks |
Country of Origin | United States |
Product Type | Calf Bovine Serum |
Sterilization Method | Sequential-filtered, 100 nm |
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Processing | Quantity | Price | Quantity & Availability | ||||
16211092
|
Cytiva
SH30072.04IH25-40 |
Heat Inactivated, Irradiated (25-40 kGy) | 1000 mL |
£157.00
1 L |
Estimated Shipment: 06-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12829447
|
Cytiva
SH30072.03IH |
Irradiated, Heat Inactivated | 500 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16685735
|
Cytiva
SH30072.04IR35 |
Irradiated | 1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16605745
|
Cytiva
SH30072.04NI |
Non-irradiated | 1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16625755
|
Cytiva
SH30372.02 |
None | 100 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16635755
|
Cytiva
SH30372.04 |
None | 1000 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Supplemented with iron and growth-promoting factors
- Age of animal at time of collection approximately 16-24 weeks
- Virus panel testing according to 9 CFR 113.53
- Low in antibodies and high in growth factors
- Sourced in the USA with complete traceability back to original source
- A cost-effective alternative to FBS
Specifications
Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities | |
United States | |
Sequential-filtered, 100 nm | |
Virus Panel Testing According to 9 CFR 113.53 | |
Supplementing Cell Culture Growth | |
Yes |
16 to 22 weeks | |
Calf Bovine Serum | |
Formula-Fed Veal Calves | |
<10 EU/mL | |
<10 mg/dL | |
HyClone™ |